Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLPS Antibody (111), PE, Novus Biologicals™
SDP

Catalog No. NB334265 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB334265 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB334265 Supplier Novus Biologicals Supplier No. NBP290233PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CLPS Monoclonal antibody specifically detects CLPS in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen CLPS
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 111
Conjugate PE
Dilution ELISA, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias colipase, colipase, pancreatic, pancreatic colipase preproprotein
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human CLPS (Uniprot#: P04118, Met1-Gln112)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1208
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.