Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLPTM1L Antibody, Novus Biologicals™
SDP

Catalog No. NBP162477 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP162477 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP162477 Supplier Novus Biologicals Supplier No. NBP162477
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

CLPTM1L Polyclonal specifically detects CLPTM1L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CLPTM1L
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q96KA5
Gene Alias cisplatin resistance related protein CRR9p, Cisplatin resistance-related protein 9, cleft lip and palate transmembrane protein 1-like protein, CLPTM1-like, CLPTM1-like protein, CRR9DKFZp761M2324, CRR9p, DKFZp666M1010, FLJ14400, FLJ32533
Gene Symbols CLPTM1L
Host Species Rabbit
Immunogen Synthetic peptides corresponding to CLPTM1L(CLPTM1-like) The peptide sequence was selected from the middle region of CLPTM1L (NP_110409). Peptide sequence KAFTYKAFNTFIDDVFAFIITMPTSHRLACFRDDVVFLVYLYQRWLYPVD.
Molecular Weight of Antigen 62 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81037
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.