Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clusterin/APOJ Antibody (SAIC-43B-8), PerCP, Novus Biologicals™
SDP

Catalog No. NB340490 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340490 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340490 Supplier Novus Biologicals Supplier No. NBP320109PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Clusterin/APOJ Monoclonal antibody specifically detects Clusterin/APOJ in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Mass Spectrometry

Specifications

Antigen Clusterin/APOJ
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Mass Spectrometry
Classification Monoclonal
Clone SAIC-43B-8
Conjugate PerCP
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Mass Spectrometry
Formulation PBS
Gene Alias 40, 40, sulfated glycoprotein 2, Aging-associated gene 4 protein, aging-associated protein 4, apo-J, APOJ, Apolipoprotein J, CLIclusterin (complement lysis inhibitor, SP-40, clusterin, Complement cytolysis inhibitor, complement lysis inhibitor, Complement-associated protein SP-40, Ku70-binding protein 1, KUB1SGP2, MGC24903, NA1/NA2, SGP-2, SP-40, sulfated glycoprotein 2, Testosterone-repressed prostate message 2, testosterone-repressed prostate message 2, apolipoprotein J), TRPM-2TRPM2
Host Species Rabbit
Immunogen Peptide ASSIIDELFQDR derived from Clusterin conjugated to KLH. Clusterin isoform 1 functions as extracellular chaperone that prevents aggregation of nonnative proteins
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Dendritic Cell Markers, Hypoxia, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 1191
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.