Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris™ CM 10
Click to view available options
Quantity:
10 mg
50 mg
Description
CM 10 is a aldehyde dehydrogenase 1A (ALDH1A) inhibitor (IC50 values are 0.64, 0.74 and 1.7 μM for ALDH1A3, ALDH1A2 and ALDH1A1, respectively). Induces a necroptotic phenotype and preferentially depletes CD133+ ovarian cancer stem cells in vitro.
Specifications
Specifications
| Quantity | 10 mg |
| Molecular Formula | C20H23N3O |
| Formula Weight | 321.42 |
| Product Type | Inhibitor |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction