Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMC1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB354276 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB354276 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB354276 Supplier Novus Biologicals Supplier No. NBP297239PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CMC1 Polyclonal antibody specifically detects CMC1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen CMC1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias C3orf68, chromosome 3 open reading frame 68, cmc1p, COX assembly mitochondrial protein homolog, COX assembly mitochondrial protein homolog (S. cerevisiae), CX9C mitochondrial protein required for full expression of COX 1, DKFZp779D0833, MGC61571, mitochondrial metallochaperone-like protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CMC1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 152100
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.