Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMP kinase Antibody (08), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB351441 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB351441 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB351441 Supplier Novus Biologicals Supplier No. NBP306539APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CMP kinase Monoclonal antibody specifically detects CMP kinase in Human samples. It is validated for Western Blot

Specifications

Antigen CMP kinase
Applications Western Blot
Classification Monoclonal
Clone 08
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias CMKEC 2.7.4.14, CMPKUMKUMP-CMP kinase, cytidine monophosphate (UMP-CMP) kinase 1, cytosolic, Cytidine monophosphate kinase, Cytidylate kinaseUMP/CMP kinase, Deoxycytidylate kinase, EC 2.7.4, UMP/CMPK, UMP-CMPK, UMPKUCK, Uridine monophosphate kinase, Uridine monophosphate/cytidine monophosphate kinase
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CMP kinase (Uniprot#: P30085; Met1-Gly196).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 51727
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.