Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMTM6 Antibody, Novus Biologicals™
SDP

Catalog No. NBP131183 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP131183 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP131183 Supplier Novus Biologicals Supplier No. NBP131183
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CMTM6 Polyclonal antibody specifically detects CMTM6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).

Specifications

Antigen CMTM6
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Alias chemokine-like factor super family 6, chemokine-like factor superfamily 6, Chemokine-like factor superfamily member 6, CKLF-like MARVEL transmembrane domain containing 6, CKLF-like MARVEL transmembrane domain-containing protein 6, CKLFSF6, FLJ20396, PRO2219
Gene Symbols CMTM6
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human CMTM6. The exact sequence is proprietary.
Molecular Weight of Antigen 20 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 54918
Test Specificity Tested in human. Predicted cross-reactivity based on sequence identity: Rat (87%), Rhesus Monkey (100%), and Chimpanzee (100%). Not yet tested in any other species.
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.