Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNBP Antibody (38F10), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N242632PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N242632PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N242632PEC7 Supplier Novus Biologicals Supplier No. NBP242632PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CNBP Monoclonal specifically detects CNBP in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CNBP
Applications Flow Cytometry
Classification Monoclonal
Clone 38F10
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CCHC-type zinc finger, nucleic acid binding protein, cellular nucleic acid binding protein, cellular nucleic acid-binding protein, DM2, erythroid differentiation-related, FLJ11631, PROMM, RNF163CNBP1ZNF9ZCCHC22, sterol regulatory element-binding protein, zinc finger protein 273, Zinc finger protein 9, zinc finger protein 9 (a cellular retroviral nucleic acid binding protein)
Gene Symbols CNBP
Host Species Mouse
Immunogen Recombinant human CNBP (1-170aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 7555.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.