Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNOT3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP121396 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP121396 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP121396 Supplier Novus Biologicals Supplier No. NBP121396
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CNOT3 Polyclonal specifically detects CNOT3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen CNOT3
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:200, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:200
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. O75175
Gene Alias CCR4-NOT transcription complex, subunit 3, KIAA0691CCR4-associated factor 3, LENG2yeast) homolog, Leukocyte receptor cluster member 2
Gene Symbols CNOT3
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 525 and 575 of human CCR4-NOT transcription complex, subunit 3 using the numbering given in entry NP_055331.1 (GeneID 4849).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4849
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.