Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNOT6 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB326947 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB326947 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB326947 Supplier Novus Biologicals Supplier No. NBP299135APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CNOT6 Polyclonal antibody specifically detects CNOT6 in Human samples. It is validated for Western Blot

Specifications

Antigen CNOT6
Applications Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias carbon catabolite repression 4 protein, Carbon catabolite repressor protein 4 homolog, CCR4 carbon catabolite repression 4-like, CCR4CCR4-NOT transcription complex subunit 6, CCR4-NOT transcription complex, subunit 6, Cytoplasmic deadenylase, EC 3.1.13.4, KIAA1194EC 3.1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human CNOT6.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57472
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.