Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CNP Propeptide Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB3739
This item is not returnable.
View return policy
Description
CNP/NPPC Monoclonal specifically detects CNP/NPPC in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| CNP/NPPC | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P23582 | |
| NPPC | |
| E. coli-derived recombinant human CNP Lys24-Cys126 Accession # P23582 | |
| 100 μg | |
| Primary | |
| Detects human CNP Propeptide in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-react with recombinant human (rh) ANP precursor or rhBNP precursor is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 347625 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CNP, CNP2, C-type natriuretic peptide, natriuretic peptide C, natriuretic peptide precursor C, NPPC | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4880 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction