Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNPase Antibody (1H10), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB360565 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360565 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360565 Supplier Novus Biologicals Supplier No. NBP250031MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CNPase Monoclonal antibody specifically detects CNPase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry Free-Floating

Specifications

Antigen CNPase
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Free Floating)
Classification Monoclonal
Clone 1H10
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry - Free Floating
Formulation 50mM Sodium Borate
Gene Alias 2', 3' cyclic nucleotide 3' phosphohydrolase, 2'-3'-cyclic nucleotide 3' phosphodiesterase, 2'-3'-cyclic-nucleotide 3'-phosphodiesterase, CNP1, CNPase, EC 3.1.4.37
Host Species Mouse
Immunogen Full length human recombinant CNPase protein expressed in and purified from E. coli. [UniProt# P09543]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Glia Markers, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1267
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.