Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNPase Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11079874 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11079874 0.1 mg
NB018029 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11079874 Supplier Novus Biologicals Supplier No. NB11079874
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CNPase Polyclonal specifically detects CNPase in Human samples. It is validated for Western Blot.

Specifications

Antigen CNPase
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Alias 2', 3' cyclic nucleotide 3' phosphohydrolase, 2'-3'-cyclic nucleotide 3' phosphodiesterase, 2'-3'-cyclic-nucleotide 3'-phosphodiesterase, CNP1, CNPase, EC 3.1.4.37
Gene Symbols CNP
Host Species Rabbit
Immunogen Reacts with residues 68-84 of the 46-48 (doublet) kDa human, CNPase protein (74% sequence identity and 94% sequence similarity to mouse and rat).
Molecular Weight of Antigen 45 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1267
Test Specificity CNPase - Oligodendrocyte Marker
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.