Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNPase Antibody, Novus Biologicals™
SDP

Catalog No. NB11093362 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11093362 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11093362 Supplier Novus Biologicals Supplier No. NB11093362
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CNPase Polyclonal specifically detects CNPase in Human, Mouse, Rat, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen CNPase
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:1000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:500-1:1000
Formulation Neat whole antisera with No Preservative
Gene Accession No. P13233
Gene Alias 2', 3' cyclic nucleotide 3' phosphohydrolase, 2'-3'-cyclic nucleotide 3' phosphodiesterase, 2'-3'-cyclic-nucleotide 3'-phosphodiesterase, CNP1, CNPase, EC 3.1.4.37
Gene Symbols CNP
Host Species Rabbit
Immunogen Endogenous rabbit 2,3 cyclic nucleotide-3-phospho-diesterase. Accession # P13233
Molecular Weight of Antigen 46 kDa
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1267
Test Specificity Specific for the ∼ 46k CNP protein in Western blots.
Target Species Human, Mouse, Rat, Rabbit, Sheep
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.