Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNPY2 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB086826 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB086826 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB086826 Supplier Novus Biologicals Supplier No. NBP300189G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CNPY2 Polyclonal specifically detects CNPY2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CNPY2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50 mM Sodium Borate
Gene Alias canopy 2 homolog (zebrafish), HP10390, MSAPprotein canopy homolog 2, Putative secreted protein Zsig9, TMEM4CNPY4, Transmembrane protein 4MIR-interacting saposin-like protein, ZSIG9Cnpy2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant CNPY2 (Q9Y2B0-1; Met1-Ser178).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10330
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.