Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNRIP1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB341959 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341959 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341959 Supplier Novus Biologicals Supplier No. NBP281897MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CNRIP1 Polyclonal antibody specifically detects CNRIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen CNRIP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C2orf32, cannabinoid receptor CB1-interacting protein 1, cannabinoid receptor interacting protein 1, CB1 cannabinoid receptor-interacting protein 1, chromosome 2 open reading frame 32, CRIP1, CRIP-1, CRIP1a, CRIP1b, DKFZp566K1924
Host Species Rabbit
Immunogen CNRIP1 antibody was raised against a 15 amino acid peptide near the center of human CNRIP1. The immunogen is located within amino acids 50 - 100 of CNRIP1. Amino Acid Squence: VPLELKSKEPDGDRV
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 25927
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.