Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNTF Antibody (121), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB078707 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB078707 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB078707 Supplier Novus Biologicals Supplier No. NBP290839AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CNTF Monoclonal specifically detects CNTF in Rat samples. It is validated for ELISA.

Specifications

Antigen CNTF
Applications ELISA
Classification Monoclonal
Clone 121
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias ciliary neurotrophic factor, HCNTF
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Rat CNTF (P20294; Met1-Met200).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1270
Test Specificity No cross-reactivity in ELISA with: Human CNTF Mouse LIF Mouse OSM Rat IL6
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.