Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNTF Antibody (4-68) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB019448 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019448 0.025 mg
NBP229742 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019448 Supplier Novus Biologicals Supplier No. NBP2297420.025MG

Mouse Monoclonal Antibody

CNTF Monoclonal specifically detects CNTF in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen CNTF
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 24929
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 10-20 ug/ml
Formulation 20mM Phosphate (pH 7.6) and 0.25M NaCl with 0.1% Sodium Azide
Gene Accession No. P26441
Gene Alias ciliary neurotrophic factor, HCNTF
Gene Symbols CNTF
Host Species Mouse
Immunogen Purified ciliary neurotrophic factor from rat sciatic nerve.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1270
Test Specificity Ciliary neurotrophic factor. Detects a 22 kD band in Western Blots of rat sciatic nerve extract.
Target Species Primate, Mouse, Human, Rat, Rabbit
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.