Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNTFR Antibody (MM0190-10L29), Biotin, Novus Biologicals™
SDP

Catalog No. NBP212196B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212196B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212196B Supplier Novus Biologicals Supplier No. NBP212196B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CNTFR Monoclonal specifically detects CNTFR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CNTFR
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0190-10L29
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias ciliary neurotrophic factor receptor, ciliary neurotrophic factor receptor alpha, ciliary neurotrophic factor receptor subunit alpha, CNTF receptor subunit alpha, CNTFR alpha, CNTFR-alpha, MGC1774
Gene Symbols CNTFR
Host Species Mouse
Immunogen Human recombinant CNTFR
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1271
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.