Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNTFR Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | CNTFR |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP16894320
![]() |
Novus Biologicals
NBP16894320UL |
20 μL |
Each for $206.00
|
|
|||||
NBP168943
![]() |
Novus Biologicals
NBP168943 |
100 μL |
Each for $487.50
|
|
|||||
Description
CNTFR Polyclonal specifically detects CNTFR in Human samples. It is validated for Western Blot.Specifications
CNTFR | |
Polyclonal | |
Rabbit | |
Growth and Development, Neuronal Cell Markers | |
ciliary neurotrophic factor receptor, ciliary neurotrophic factor receptor alpha, ciliary neurotrophic factor receptor subunit alpha, CNTF receptor subunit alpha, CNTFR alpha, CNTFR-alpha, MGC1774 | |
CNTFR | |
IgG | |
This product is specific to Subunit or Isoform: alpha. |
Western Blot | |
Unconjugated | |
RUO | |
P26992 | |
1271 | |
Synthetic peptides corresponding to CNTFR (ciliary neurotrophic factor receptor) The peptide sequence was selected from the C terminal of CNTFR. Peptide sequence VAAHATPWTEEPRHLTTEAQAAETTTSTTSSLAPPPTTKICDPGELGSGG. | |
Primary | |
41 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title