Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CNTNAP5 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5122083
This item is not returnable.
View return policy
Description
May play a role in the correct development and proper functioning of the peripheral and central nervous system and be involved in cell adhesion and intercellular communication.
Specifications
CNTNAP5 | |
Polyclonal | |
Unconjugated | |
CNTNAP5 | |
CASPR5; cell recognition molecule Caspr5; CNTNAP5; contactin associated protein like 5; contactin associated protein-like 5; contactin-associated protein-like 5 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
129684 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q8WYK1 | |
CNTNAP5 | |
Ala25-Asp302. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction