Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNTNAP5 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | CNTNAP5 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5122084
|
Invitrogen™
PA5122084 |
20 μL |
Each for $209.50
|
|
|||||
PIPA5122083
![]() |
Invitrogen™
PA5122083 |
100 μL |
Each for $511.50
|
|
|||||
Description
May play a role in the correct development and proper functioning of the peripheral and central nervous system and be involved in cell adhesion and intercellular communication.Specifications
CNTNAP5 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q8WYK1 | |
129684 | |
Ala25-Asp302. | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
CNTNAP5 |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
CASPR5; cell recognition molecule Caspr5; CNTNAP5; contactin associated protein like 5; contactin associated protein-like 5; contactin-associated protein-like 5 | |
CNTNAP5 | |
IgG | |
Antigen affinity chromatography, Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title