Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Co 101244 hydrochloride
GREENER_CHOICE

Catalog No. 245610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-561-0 10 mg
24-565-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 24-561-0 Supplier Tocris Bioscience™ Supplier No. 2456/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

GluN2B-selective NMDA antagonist

Novel, potent and selective antagonist of GluN2B (formally NR2B)-containing NMDA receptors (IC50 values are 0.043, > 100 and > 100 μM for GluN1A/2B (NR1A/2B), GluN1A/2A (NR1A/2A) and GluN1A/2C (NR1A/2C) subunit combinations respectively). Displays neuroprotective effects in vivo and in vitro.

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Chemical Identifiers

CAS 193356-17-1
Molecular Formula C21H28ClNO3

Specifications

Quantity 10 mg
Formula Weight 377.91
Purity >99%
Molecular Weight (g/mol) 377.909
Chemical Name or Material Co 101244 hydrochloride
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.