Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COA6 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB135733 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB135733 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB135733 Supplier Novus Biologicals Supplier No. NBP306320AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

COA6 Polyclonal specifically detects COA6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen COA6
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias chromosome 1 open reading frame 31, cytochrome c oxidase assembly factor 6 homolog (S. cerevisiae), hypothetical protein LOC388753
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human COA6 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388753
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.