Learn More
Coactosin-like Protein 1/CotL1 Antibody (1D6) - BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Coactosin-like Protein 1/CotL1 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 1D6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, ELISA 1:1000 |
| Formulation | PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide |
| Gene Alias | CLPFLJ43657, coactosin-like 1 (Dictyostelium), coactosin-like protein, MGC19733 |
| Gene Symbols | COTL1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.