Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coactosin-like Protein 1/CotL1 Antibody (1D6) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP23120305 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP23120305 0.05 mL
NBP23120301 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP23120305 Supplier Novus Biologicals Supplier No. NBP2312030.05ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Coactosin-like Protein 1/CotL1 Monoclonal specifically detects Coactosin-like Protein 1/CotL1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen Coactosin-like Protein 1/CotL1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1D6
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:1000
Formulation PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide
Gene Alias CLPFLJ43657, coactosin-like 1 (Dictyostelium), coactosin-like protein, MGC19733
Gene Symbols COTL1
Host Species Mouse
Immunogen Recombinant human COTL1 (1-142aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.05 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23406
Test Specificity Anti-human COTL1 mAb, clone 1D6, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human COTL1 protein.
Reconstitution Protein A purified
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.