Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coagulation Factor II/Thrombin Antibody (318), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB332786 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB332786 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB332786 Supplier Novus Biologicals Supplier No. NBP290543MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Coagulation Factor II/Thrombin Monoclonal antibody specifically detects Coagulation Factor II/Thrombin in Mouse samples. It is validated for Western Blot

Specifications

Antigen Coagulation Factor II/Thrombin
Applications Western Blot
Classification Monoclonal
Clone 318
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Coagulation Factor II, coagulation factor II (thrombin) receptor-like 2, Coagulation factor II receptor-like 2, Coagulation factor II receptor-like 2 (protease-actovated receptor 3), PAR-3, PAR3proteinase-activated receptor 3, protease-activated receptor 3, proteinase-activated receptor-3, PT, serine protease, Thrombin receptor-like 2
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse Coagulation Factor II/Thrombin (Accession#: NP_034298.1, Met1-Gly618)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2147
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.