Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coagulation Factor III/Tissue Factor Antibody (HTF-1), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB002260 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002260 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002260 Supplier Novus Biologicals Supplier No. NBP262199UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Coagulation Factor III/Tissue Factor Monoclonal specifically detects Coagulation Factor III/Tissue Factor in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry.

Specifications

Antigen Coagulation Factor III/Tissue Factor
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone HTF-1
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), FLJ17960, TF, TFA, Thromboplastin, tissue factor
Gene Symbols F3
Host Species Mouse
Immunogen Human brain tissue factor (CD142)
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2152
Test Specificity The antibody MEM-25 reacts with CD11a (alpha subunit of human LFA-1), a 170-180 kDa type I transmembrane glycoprotein expressed on B and T lymphocytes, monocytes, macrophages, neutrophils, basophils and eosinophils.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.