Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coagulation Factor III/Tissue Factor Antibody (SN20-16), Novus Biologicals™
SDP

Catalog No. NB170353 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
25 ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170353 25 ul
NBP267731 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB170353 Supplier Novus Biologicals Supplier No. NBP26773125UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Coagulation Factor III/Tissue Factor Monoclonal specifically detects Coagulation Factor III/Tissue Factor in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen CoagulationFactorIII/TissueFactor
Applications Immunofluorescence
Classification Monoclonal
Clone SN20-16
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:500
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), FLJ17960, TF, TFA, Thromboplastin, tissue factor
Gene Symbols F3
Host Species Rabbit
Immunogen Synthetic peptide within human Coagulation Factor III/Tissue Factor aa 30-70. (SwissProt: P13726 Human; SwissProt: P20352 Mouse; SwissProt: P42533 Rat)
Purification Method Protein A purified
Quantity 25 ul
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2152
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.