Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coagulation Factor VII Antibody (121), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB072659 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB072659 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB072659 Supplier Novus Biologicals Supplier No. NBP290369AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Coagulation Factor VII Monoclonal specifically detects Coagulation Factor VII in Mouse samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen Coagulation Factor VII
Applications ELISA
Classification Monoclonal
Clone 121
Conjugate Alexa Fluor 594
Dilution ELISA, Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias coagulation factor VII, coagulation factor VII (serum prothrombin conversion accelerator), EC 3.4.21, EC 3.4.21.21, Eptacog alfa, FVII coagulation protein, Proconvertin, Serum prothrombin conversion accelerator, SPCA
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Coagulation Factor VII (NP_034302.2; Met 1-Leu 446).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 2155
Test Specificity No cross-reactivity in ELISA with: Human F10/ FX/ Factor X Human F11/ FXI/ Factor XI Human Kallikrein-1/ KLK-1 Human Kallikrein-13/ KLK-13 Human Kallikrein-11/ KLK-11 Human Kallikrein-7/ KLK-7
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.