Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coagulation Factor VII Antibody (321605), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB365524 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB365524 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB365524 Supplier Novus Biologicals Supplier No. FAB23381K
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Coagulation Factor VII Monoclonal antibody specifically detects Coagulation Factor VII in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Coagulation Factor VII
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 321605
Conjugate DyLight 488
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias coagulation factor VII, coagulation factor VII (serum prothrombin conversion accelerator), EC 3.4.21, EC 3.4.21.21, Eptacog alfa, FVII coagulation protein, Proconvertin, Serum prothrombin conversion accelerator, SPCA
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor VII, Ala39-Pro444, Accession # NP_062562
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 2155
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.