Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coagulation Factor VII Rabbit anti-Mouse, Clone: 121, Novus Biologicals™
SDP

Catalog No. NB022192 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB022192 50 μg
NB022191 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB022192 Supplier Novus Biologicals Supplier No. NBP29036950UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Coagulation Factor VII Monoclonal specifically detects Coagulation Factor VII in Mouse samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen Coagulation Factor VII
Applications ELISA
Classification Monoclonal
Clone 121
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias coagulation factor VII, coagulation factor VII (serum prothrombin conversion accelerator), EC 3.4.21, EC 3.4.21.21, Eptacog alfa, FVII coagulation protein, Proconvertin, Serum prothrombin conversion accelerator, SPCA
Host Species Rabbit
Immunogen Recombinant Mouse Coagulation Factor VII protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2155
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.