Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coagulation Factor X Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB089038 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB089038 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB089038 Supplier Novus Biologicals Supplier No. NBP299698AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Coagulation Factor X Polyclonal specifically detects Coagulation Factor X in Human samples. It is validated for ELISA.

Specifications

Antigen Coagulation Factor X
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias coagulation factor X, EC 3.4.21, EC 3.4.21.6, factor Xa, FX, FXA, prothrombinase, Stuart factor, Stuart-Prower factor
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Coagulation Factor X (NP_000495.1; Met 1-Lys 488).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2159
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.