Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coagulation Factor X Antibody (MM0192-10A10), PerCP, Novus Biologicals™
SDP

Catalog No. NB361274 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361274 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361274 Supplier Novus Biologicals Supplier No. NBP212198PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Coagulation Factor X Monoclonal antibody specifically detects Coagulation Factor X in Human samples. It is validated for Western Blot

Specifications

Antigen Coagulation Factor X
Applications Western Blot
Classification Monoclonal
Clone MM0192-10A10
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias coagulation factor X, EC 3.4.21, EC 3.4.21.6, factor Xa, FX, FXA, prothrombinase, Stuart factor, Stuart-Prower factor
Host Species Mouse
Immunogen Human recombinant Coagulation Factor X
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2159
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.