Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coagulation Factor XI Antibody (MM0193-7C38), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP212199IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212199IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212199IR Supplier Novus Biologicals Supplier No. NBP212199IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Coagulation Factor XI Monoclonal specifically detects Coagulation Factor XI in Human samples. It is validated for Western Blot.

Specifications

Antigen Coagulation Factor XI
Applications Western Blot
Classification Monoclonal
Clone MM0193-7C38
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias coagulation factor XI, EC 3.4.21, EC 3.4.21.27, FXIPlasma thromboplastin antecedent, MGC141891, PTA
Gene Symbols F11
Host Species Mouse
Immunogen Human recombinant Coagulation Factor XI
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2160
Test Specificity Detects human Coagulation factor XI (Light Chain)
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.