Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coagulation Factor XIV/Protein C Antibody (PC 7), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP10113A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP10113A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP10113A94 Supplier Novus Biologicals Supplier No. NBP105113AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Coagulation Factor XIV/Protein C Monoclonal specifically detects Coagulation Factor XIV/Protein C in Human samples. It is validated for ELISA.

Specifications

Antigen Coagulation Factor XIV/Protein C
Applications ELISA
Classification Monoclonal
Clone PC 7
Conjugate Alexa Fluor 594
Dilution ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Anticoagulant protein C, APC, Autoprothrombin IIA, Blood coagulation factor XIV, EC 3.4.21, EC 3.4.21.69, PC, PROC1, protein C (inactivator of coagulation factors Va and VIIIa), vitamin K-dependent protein C
Gene Symbols PROC
Host Species Mouse
Immunogen Recombinant form of human activated protein C (Drotrecogin alfa, Xigris)
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5624
Test Specificity Has specificity for a conformation dependent neoepitope that is expressed in activated protein C upon complex-formation with alpha1-antitrypsin. No reaction is seen to non-complexed alpha1-antitrypsin and only very little cross reaction to protein C zymogen. Note that specificity is calcium dependent.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.