Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coagulation Factor XIV/Protein C Antibody (PC 7), PerCP, Novus Biologicals™
SDP

Catalog No. NB359058 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359058 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359058 Supplier Novus Biologicals Supplier No. NBP105113PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Coagulation Factor XIV/Protein C Monoclonal antibody specifically detects Coagulation Factor XIV/Protein C in Human samples. It is validated for ELISA

Specifications

Antigen Coagulation Factor XIV/Protein C
Applications ELISA
Classification Monoclonal
Clone PC 7
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias Anticoagulant protein C, APC, Autoprothrombin IIA, Blood coagulation factor XIV, EC 3.4.21, EC 3.4.21.69, PC, PROC1, protein C (inactivator of coagulation factors Va and VIIIa), vitamin K-dependent protein C
Host Species Mouse
Immunogen Recombinant human activated protein C
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5624
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.