Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coagulation Factor XIV/Protein C Rabbit anti-Human, Mouse, Rat, Clone: 3W3R7, Novus Biologicals™
SDP

Catalog No. NB167945 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167945 100 μg
NB167946 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167945 Supplier Novus Biologicals Supplier No. NBP316427100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Coagulation Factor XIV/Protein C Monoclonal antibody specifically detects Coagulation Factor XIV/Protein C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence

Specifications

Antigen Coagulation Factor XIV/Protein C
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 3W3R7
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Anticoagulant protein C, APC, Autoprothrombin IIA, Blood coagulation factor XIV, EC 3.4.21, EC 3.4.21.69, PC, PROC1, protein C (inactivator of coagulation factors Va and VIIIa), vitamin K-dependent protein C
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 300-461 of human Coagulation Coagulation Factor XIV/Protein C (P04070).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5624
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.