Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Coarse White Marble Specimen (Metamorphic Rock)

Click to view available options
Quantity:
12/Pk.
Each
Description
- For use in classroom identification labs and exercises
- Correlates well to Next Generation Science Standards Evidence Statements
- For use in classroom activities trying to ascertain differences between metamorphic rocks and their provenance
- Denser than limestone, yet effervesce in acid
- Comes in clear labeled and barcoded bag for storage
Specifications
Specifications
| Quantity | 12/Pk. |
| For Use With (Application) | Classroom activities to ascertain differences between metamorphic rocks and their provenance |
| Product Type | Coarse White Marble Study Specimen (Metamorphic Rock) |
| Width (English) | Approx. 1 in. |
| Width (Metric) | Approx. 3 cm |
| Packaging | Clear labeled and barcoded bag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction