Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COASY Antibody (OTI3B3), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N272224A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272224A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272224A405 Supplier Novus Biologicals Supplier No. NBP272224AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

COASY Monoclonal specifically detects COASY in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen COASY
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3B3
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias bifunctional phosphopantetheine adenylyl transferase/dephospho CoA kinase, CoA synthaseUKR1, CoASY, Coenzyme A synthase, DPCK, FLJ35179, NBPbifunctional coenzyme A synthase, nucleotide binding protein, phosphopantetheine adenylyltransferase/ dephosphocoenzyme A kinase, POV-2, PPAT
Gene Symbols COASY
Host Species Mouse
Immunogen Full length recombinant protein of human COASY (NP_079509) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 80347.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.