Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Magnesium cation (3 x 30 ul)
Click to view available options
Quantity:
3 x 30 μL
Description
- 3 vials of 30μL
- For optimizing TdT labeling in TACS in situ apoptosis detection kits.
- Magnesium is useful for tissue that demonstrates high background.
Specifications
Specifications
Quantity | 3 x 30 μL |
Molecular Formula | MG2+ |
Product Type | Magnesium cation |
Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction