Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cobalt foil, 1.0mm (0.04 in.) thick, Puratronic™, 99.995% (metals basis), Thermo Scientific™
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Specifications
Quantity | 50 x 50 mm |
Odor | Odorless |
Weight | ≈5.6g/25x25mm |
Form | Foil |
Percent Purity | 99.995% |
Grade | Puratronic™ |
Assay | (metals basis) |
Chemical Name or Material | Cobalt foil, 1.0mm (0.04 in.) thick |
Safety and Handling
missing translation for 'healthHazard1' | H317-H334-H350 |
missing translation for 'healthHazard2' | GHS H Statement H334-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. |
missing translation for 'healthHazard3' | P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c |
EINECSNumber | 231-158-0 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction