Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxylamine-O-sulfonic acid, 97%

Catalog No. AC325940250
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle

Catalog No. AC325940250

Supplier: Thermo Scientific Chemicals 325940250

Only null left
Add to Cart

CAS: 2950-43-8 | H3NO4S | 113.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2950-43-8
Molecular Formula H3NO4S
Molecular Weight (g/mol) 113.09
MDL Number MFCD00011604
InChI Key DQPBABKTKYNPMH-UHFFFAOYSA-N
Synonym hydroxylamine-o-sulfonic acid, aminooxy sulfonic acid, sulfoperamidic acid, hydroxylamine-o-sulphonic acid, permonosulfamic acid, amidoperoxymonosulfuric acid, aminooxysulfonic acid, amidosulfonic peracid, haos, sulfamic acid n-oxide
PubChem CID 76284
IUPAC Name amino hydrogen sulfate
SMILES NOS(O)(=O)=O

Specifications

Melting Point 210.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 0.97
Packaging Glass bottle
Linear Formula H2NOSO3H
Quantity 25 g
Fieser 01,481; 02,217; 03,156; 04,256; 05,343; 06,290; 08,250; 09,245; 10,207; 15,17
Solubility Information Solubility in water: soluble
Formula Weight 113.09
Percent Purity 0.97
Physical Form Crystalline Powder
Chemical Name or Material Hydroxylamine-O-sulfonic acid, 0.97
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Suspected of causing cancer.
Causes serious eye damage.
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Use personal protective equipment as required.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 220-971-6
RTECSNumber NC5427000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.