Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) nitrate hexahydrate, 97.7% min

Catalog No. p-7048722
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg

CAS: 10026-22-9 | CoH12N2O12 | 291.03 g/mol

Cobalt(II) nitrate hexahydrate is used in the preparation of metallic cobalt and cobalt based dyes and pigments. It is a precursor for the preparation of catalyst for use in certain organic reactions. It finds use in catalysis, vitamin preparations, and porcelain decoration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-22-9
Molecular Formula CoH12N2O12
Molecular Weight (g/mol) 291.03
MDL Number MFCD00149647
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
Synonym cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
PubChem CID 24821
ChEBI CHEBI:86214
IUPAC Name cobalt(2+);dinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 55°C to 56°C
Color Red
Physical Form Crystalline
Quantity 25 g
UN Number UN1477
Merck Index 14,2444
Solubility Information Soluble in water,alcohol,acetone and ammonia.
Formula Weight 291.04 (182.94 Anhydrous)
Percent Purity ≤97.7%
Odor Odorless
Refractive Index 1.52
Density 1.87 g/mL
Chemical Name or Material Cobalt(II) nitrate hexahydrate
Show More Show Less
Hazard Category H272-H302+H332-H317-H318-H334-H341-H350i-H360F
Hazard Statement GHS H Statement
H272-H334-H350-H360-H341-H302-H317
May intensify fire; oxidizer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P220-P221-P261-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P342+P311-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
EINECSNumber 233-402-1
RTECSNumber QU7355500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.