Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COBLL1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB319711 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB319711 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB319711 Supplier Novus Biologicals Supplier No. NBP298671MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

COBLL1 Polyclonal antibody specifically detects COBLL1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen COBLL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias COBL-like 1, COBLR1, cordon-bleu protein-like 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human COBLL1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22837
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.