Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COBRA1 Antibody (F7E4), Novus Biologicals™
SDP

Catalog No. NBP229529 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229529 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP229529 Supplier Novus Biologicals Supplier No. NBP229529
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

COBRA1 Monoclonal specifically detects COBRA1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen COBRA1
Applications Western Blot
Classification Monoclonal
Clone F7E4
Conjugate Unconjugated
Dilution Western Blot 1:1000
Gene Accession No. Q8WX92
Gene Alias cofactor of BRCA1DKFZp586B0519, KIAA1182negative elongation factor B, NELFB, NELF-Bnegative elongation factor protein B
Gene Symbols COBRA1
Host Species Mouse
Immunogen Recombinant full length human COBRA1 purified from E. coli. Accession # Q8WX92
Molecular Weight of Antigen 62 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 25920
Test Specificity Specific for the ∼62k COBRA1 protein in Western blots of neonatal rat brain lysate.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.