Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cochlin Antibody, Novus Biologicals™
SDP

Catalog No. NB6914120UL Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6914120UL 20 μL
NBP169141 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB6914120UL Supplier Novus Biologicals Supplier No. NBP16914120UL

Rabbit Polyclonal Antibody

Cochlin Polyclonal specifically detects Cochlin in Human samples. It is validated for Western Blot.

Specifications

Antigen Cochlin
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. O43405
Gene Alias coagulation factor C (Limulus polyphemus homolog); cochlin, coagulation factor C homolog, cochlin (Limulus polyphemus), COCH-5B2COCH5B2, cochlin, DFNA9
Gene Symbols COCH
Host Species Rabbit
Immunogen Synthetic peptides corresponding to COCH (coagulation factor C homolog, cochlin (Limulus polyphemus)) The peptide sequence was selected from the C terminal of COCH. Peptide sequence VAWAPLDDLKDMASKPKESHAFFTREFTGLEPIVSDVIRGICRDFLESQQ.
Molecular Weight of Antigen 57 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1690
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.