Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COCO/DAND5 Antibody (OTI3D4), HRP, Novus Biologicals™
SDP

Catalog No. NBP272457H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272457H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272457H Supplier Novus Biologicals Supplier No. NBP272457H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

COCO/DAND5 Monoclonal specifically detects COCO/DAND5 in Human samples. It is validated for Western Blot.

Specifications

Antigen COCO/DAND5
Applications Western Blot
Classification Monoclonal
Clone OTI3D4
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias CER2CRL2, cerberus-like 2, Cerberus-like protein 2, cerl-2, CERL2, CKTSF1B3MGC126849, COCO, Cysteine knot superfamily 1, BMP antagonist 3, DAN domain family member 5, DAN domain family, member 5, DANTE, DTE, FLJ38607, GREM3cerberus 2, Gremlin-3, SP1
Gene Symbols DAND5
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 23-189 of human COCO/DAND5 (NP_689867) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 199699.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.