Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cofilin Antibody, Novus Biologicals™
SDP

Catalog No. NB1001900 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001900 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001900 Supplier Novus Biologicals Supplier No. NB1001900
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Cofilin Polyclonal specifically detects Cofilin in Canine, Human, Mouse, Pig, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Microarray, Simple Western, Western Blot.

Specifications

Antigen Cofilin
Applications Immunocytochemistry, Immunofluorescence, Microarray, Simple Western, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:1000
Formulation 10mM PBS (pH 7.4) with 15mM Sodium Azide
Gene Accession No. P23528
Gene Alias 18 kDa phosphoprotein, CFL, cofilin 1 (non-muscle), Cofilin, non-muscle isoform, cofilin-1, p18
Gene Symbols CFL1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 154-166 of human cofilin with N-terminal cysteine added, conjugated to maleimide-activated keyhole limpet hemocyanin (KLH). The corresponding immunizing sequence is identical in pig and rat non-muscle cofilin and differs by three amino acids from that of human and chicken muscle cofilin.
Purification Method IgG purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 1072
Test Specificity Detects a band at approximately 19kDa. Reacts with muscle and non-muscle cofilin.
Target Species Canine, Human, Rat, Mouse, Pig, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.