Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cofilin, p Ser3 Antibody, Novus Biologicals™
SDP

Catalog No. NB259853 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259853 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB259853 Supplier Novus Biologicals Supplier No. NBP323231
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cofilin Polyclonal antibody specifically detects Cofilin in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen Cofilin
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:50
Formulation PBS, 1 mg/ml BSA and 50% glycerol
Gene Alias 18 kDa phosphoprotein, CFL, cofilin 1 (non-muscle), Cofilin, non-muscle isoform, cofilin-1, p18
Host Species Rabbit
Immunogen A synthetic phospho-peptide (coupled to KLH) corresponding to amino acid residues surrounding serine 3 in human Cofilin. This sequence has 100% homology with similar regions of rat and mouse Cofilin, and has two amino acid differences from human Cofilin 2. [UniProt# P23528]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Neuronal Cell Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1072
Target Species Human, Mouse, Rat, Chicken
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.